Organic analytical standards, (±)-Nicotine-d3 salicylate, neat, reference material, Honeywell Fluka™
HONCR1495-10MG
:
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:1173021-00-5
- Description:(±)-Nicotine-d₃ salicylate
- Formula:C₁₇H₁₇D₃N₂O₃
- Storage temperature:Ambient
- Description:(±)-Nicotine-d₃ salicylate
- Freezing point:110...111 °C
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results