Pharmaceutical analytical standard, A1-Aminohydantoin (AHD), reference material, Honeywell Fluka™
Catalog # HONCR1443-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:6301-02-6
- Description:A1-Aminohydantoin (AHD)
- Formula:C₃H₅N₃O₂
- Storage temperature:Ambient
- Description:A1-Aminohydantoin (AHD)
- Freezing point:191...193 °C
Specifications
About this item
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results