Order Entry
Czech Republic
ContactUsLinkComponent
Pharmaceutical analytical standard, 4-Methylaminoantipyrine-d3 picrate, reference material, Honeywell Fluka™
Pharmaceutical analytical standard, 4-Methylaminoantipyrine-d3 picrate, reference material, Honeywell Fluka™
Catalog # HONCR1422-10MG
CAS Number:  
Pharmaceutical analytical standard, 4-Methylaminoantipyrine-d3 picrate, reference material, Honeywell Fluka™
Catalog # HONCR1422-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • Description:
    4-Methylaminoantipyrine-d₃ picrate
  • Formula:
    C12H12D3N3O · C6H3N3O7
  • Storage temperature:
    Ambient
  • Description:
    4-Methylaminoantipyrine-d₃ picrate
  • Freezing point:
    170...172 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results