Pharmaceutical analytical standard, Salmeterol xinafoate, reference material, Honeywell Fluka™
Katalogové číslo HONCR1415-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:94749-08-3
- Description:Salmeterol xinafoate
- Formula:C₂₅H₃₇NO₄ · C₁₁H₈O₃
- Storage temperature:Ambient
- Description:Salmeterol xinafoate
- Freezing point:116...118 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results