Pharmaceutical analytical standard, (R,R)-Formoterol tartrate, reference material, Honeywell Fluka™
Catalog # HONCR1410-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:200815-49-2
- Description:(R,R)-Formoterol tartrate
- Formula:C₁₉H₂₄N₂O₄
- Storage temperature:Ambient
- Description:(R,R)-Formoterol tartrate
- Freezing point:183...190 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results