Order Entry
Czech Republic
ContactUsLinkComponent
Pharmaceutical analytical standard, Mebendazole-amine-13C6, reference material, Honeywell Fluka™
Pharmaceutical analytical standard, Mebendazole-amine-13C6, reference material, Honeywell Fluka™
Catalog # HONCR1399-10MG
CAS Number:  
Pharmaceutical analytical standard, Mebendazole-amine-13C6, reference material, Honeywell Fluka™
Catalog # HONCR1399-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • Description:
    Mebendazole-amine-¹³C₆
  • Formula:
    C₈¹³C₆H₁₁N₃O
  • Storage temperature:
    Ambient
  • Description:
    Mebendazole-amine-¹³C₆
  • Freezing point:
    188...190 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results