Pharmaceutical analytical standard, rac-Monepantel, reference material, Honeywell Fluka™
HONCR1396-10MG
:
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:851976-50-6
- Description:rac-Monepantel
- Formula:C₂₀H₁₃F₆N₃O₂S
- Storage temperature:2...8 °C
- Description:rac-Monepantel
- Freezing point:163...164 °C
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results