Order Entry
Czech Republic
ContactUsLinkComponent
Pesticide reference standard, Amiprofos methyl, neat, Honeywell Fluka™
Pesticide reference standard, Amiprofos methyl, neat, Honeywell Fluka™
Catalog # HONCR1311-50MG
CAS Number:  
Pesticide reference standard, Amiprofos methyl, neat, Honeywell Fluka™
Catalog # HONCR1311-50MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mg
  • CAS No.:
    36001-88-4
  • Description:
    Amiprofos methyl
  • Formula:
    C₁₁H₁₇N₂O₄PS
  • Storage temperature:
    2...8 °C
  • Description:
    Amiprofos methyl
  • Freezing point:
    62...68 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results