Order Entry
Czech Republic
ContactUsLinkComponent
Pesticide reference standard, Flusulfamide, neat, Honeywell Fluka™

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mg
  • CAS No.:
    106917-52-6
  • Description:
    Flusulfamide
  • Environmentally Preferable:
  • Formula:
    C₁₃H₇Cl₂F₃N₂O₄S
  • Storage temperature:
    2...8 °C
  • Description:
    Flusulfamide
  • Freezing point:
    182...187 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results