Order Entry
Czech Republic
ContactUsLinkComponent
 
Pharmaceutical analytical standard, Sulfisoxazole-(phenyl-13C6), reference material, Honeywell Fluka™
  HONCR1246-10MG
 :  
Pharmaceutical analytical standard, Sulfisoxazole-(phenyl-13C6), reference material, Honeywell Fluka™
  HONCR1246-10MG
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • CAS No.:
    1334378-46-9
  • Description:
    Sulfisoxazole-(phenyl-¹³C₆)
  • Formula:
    ¹³C₆C₅H₁₃N₃O₃S
  • Storage temperature:
    Ambient
  • Description:
    Sulfisoxazole-(phenyl-¹³C₆)
  • Freezing point:
    192 °C

 

 

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results