Pharmaceutical analytical standard, Salbutamol acetate, reference material, Honeywell Fluka™
Catalog # HONCR1212-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:1420043-41-9
- Description:Salbutamol acetate
- Formula:C₁₃H₂₁NO₃ · C₂H₄O₂
- Storage temperature:Ambient
- Description:Salbutamol acetate
- Freezing point:158...162 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results