Order Entry
Czech Republic
ContactUsLinkComponent
Pharmaceutical analytical standard, Orciprenaline-(isopropyl-d7) acetate salt hemihydrate, reference material, Honeywell Fluka™
Pharmaceutical analytical standard, Orciprenaline-(isopropyl-d7) acetate salt hemihydrate, reference material, Honeywell Fluka™
Catalog # HONCR1154-10MG
CAS Number:  
Pharmaceutical analytical standard, Orciprenaline-(isopropyl-d7) acetate salt hemihydrate, reference material, Honeywell Fluka™
Catalog # HONCR1154-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • Description:
    Orciprenaline-(isopropyl-d₇) acetate salt hemihydrate
  • Formula:
    C11D7H10NO3 · C2H4O2 · 0.5H2O
  • Storage temperature:
    Ambient
  • Description:
    Orciprenaline-(isopropyl-d₇) acetate salt hemihydrate
  • Freezing point:
    74 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results