Pharmaceutical analytical standard, Pirbuterol-(tert-butyl-d9) acetate, reference material, Honeywell Fluka™
Catalog # HONCR1144-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:1431291-46-1
- Description:Pirbuterol-(tert-butyl-d₉) acetate
- Formula:C₁₂D₉H₁₁N₂O₃ · C2H4O2
- Storage temperature:Ambient
- Description:Pirbuterol-(tert-butyl-d₉) acetate
- Freezing point:132...136 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results