Order Entry
Czech Republic
ContactUsLinkComponent
Pesticide reference standard, Aminocyclopyrachlor, neat, Honeywell Fluka™
Pesticide reference standard, Aminocyclopyrachlor, neat, Honeywell Fluka™
  HONCR1117-25MG
 :  
Pesticide reference standard, Aminocyclopyrachlor, neat, Honeywell Fluka™
  HONCR1117-25MG
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mg
  • CAS No.:
    858956-08-8
  • Description:
    Aminocyclopyrachlor
  • Formula:
    C₈H₈ClN₃O₂
  • Storage temperature:
    Ambient
  • Description:
    Aminocyclopyrachlor

 

 

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results