Order Entry
Czech Republic
ContactUsLinkComponent
 
Pharmaceutical analytical standard, Triflupromazine-(dimethyl-d6) hydrochloride, reference material, Honeywell Fluka™
  HONCR1104-10MG
 :  
Pharmaceutical analytical standard, Triflupromazine-(dimethyl-d6) hydrochloride, reference material, Honeywell Fluka™
  HONCR1104-10MG
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • CAS No.:
    1416711-36-8
  • Description:
    Triflupromazine-(dimethyl-d₆) hydrochloride
  • Formula:
    C₁₈D₆H₁₃F₃N₂S · HCl
  • Storage temperature:
    Ambient
  • Description:
    Triflupromazine-(dimethyl-d₆) hydrochloride
  • Freezing point:
    174 °C

 

 

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results