Pharmaceutical analytical standard, Clinafloxacin, reference material, Honeywell Fluka™
Catalog # HONCR1079-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mg
- CAS No.:105956-97-6
- Description:Clinafloxacin
- Formula:C₁₇H₁₇ClFN₃O₃
- Storage temperature:2...8 °C
- Description:Clinafloxacin
- Freezing point:237...241 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results