VISOCOLOR® Colorimetric Test, Nitrite
Supplier: MACHEREY-NAGEL
Certificates
About this item
Reagents for the determination of nitrite in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Specifications
- Model:MANA931244
- Platform:Colorimetric test kits
- Storage temperature:15…25 °C
- Shelf life (months):18
- Weight:113,8 g (including packaging)
Specifications
Frequently Bought Together