ContactUsLinkComponent
Macron Fine Chemicals™ StandARd®, Platinum Cobalt Colour Standard (APHA 500)
Macron Fine Chemicals™ StandARd®, Platinum Cobalt Colour Standard (APHA 500)
Catalog # H296-03
CAS Number:  
undefined
Macron Fine Chemicals™ StandARd®, Platinum Cobalt Colour Standard (APHA 500)
Catalog # H296-03
CAS Number:  
Specifications
  • Pack type:
    Glass bottle, amber, narrow mouth
  • Pk:
    500 mL
  • Description:
    Platinum Cobalt (Pt-Co) colour standard, APHA 500
  • Seq:
    02
  • Environmentally Preferable:
Specifications
About this item
Corrosive.
APHA 500 contains 0,13% chloroplatinic acid, 0,11% cobalt (II) chloride, and 4,6% hydrogen chloride in water.