Order Entry
Czech Republic
ContactUsLinkComponent
 
Smoke point standard ASTM D 1322 / IP 57Supelco®
Catalog # SIALCRMUSPKR-250ML
Supplier:  Merck
CAS Number:  
Smoke point standard ASTM D 1322 / IP 57Supelco®
Catalog # SIALCRMUSPKR-250ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    24.25 mm
  • Pk:
    250 mL
  • Description:
    Smoke point standard
  • Test method:
    ASTM D 1322; IP 57

Specifications

About this item