Order Entry
Czech Republic
ContactUsLinkComponent
Toluene ≥99.5% ACS, Riedel-de Haën™
Irritant
Health
Flammable
Toluene ≥99.5% ACS, Riedel-de Haën™
Catalog # 179418-5L
CAS Number:  
undefined
Irritant
Health
Flammable
Toluene ≥99.5% ACS, Riedel-de Haën™
Catalog # 179418-5L
CAS Number:  

Specifications

  • Pack type:
    Aluminium bottle
  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Vapor Density:
    3.2 (vs air)
  • Assay (GC):
    Min. 99.50 %
  • Non-volatile matter:
    Max. 0.001 %
  • Water (Karl Fischer):
    Max. 0.03 %
  • S-compounds (as S):
    Max. 0.003 %
  • Reaction against H₂SO₄:
    complying
  • APHA:
    Max. 10
  • IR spectrum:
    complying
  • Vapor Pressure:
    26 mmHg ( 25 °C)

Specification Test Results

About this item

 :
 :92.14 g/mol
Merck Index:13,09607
Boiling Pt:110.6 °C
Melting Pt:-95 °C
Melting Pt:–95 °C
Density:0.867 g/cm³ (20 °C)
Flash Pt:4 °C
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
REACH:05-2114615130-69
 :GHS07
 :GHS08
 :GHS02
CAS Number:108-88-3
Formula:C₆H₅CH₃
MDL Number:MFCD00008512