Order Entry
Czech Republic
ContactUsLinkComponent
Sodium nitrite 0.2 M NaNO2 (0.2N) volumetric solution, Fluka™
Sodium nitrite 0.2 M NaNO2 (0.2N) volumetric solution, Fluka™
Catalog # 35272-1L
CAS Number:  
Sodium nitrite 0.2 M NaNO2 (0.2N) volumetric solution, Fluka™
Catalog # 35272-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002

Specification Test Results

About this item

 :
 :69 g/mol
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
CAS Number:7632-00-0
Formula:NaNO₂
MDL Number:MFCD00011118