Order Entry
Czech Republic
ContactUsLinkComponent
 
Silver nitrate 0.1 M AgNO3 (0.1N) volumetric solution, Fluka™
  72271-1L
 :  
undefined
Silver nitrate 0.1 M AgNO3 (0.1N) volumetric solution, Fluka™
  72271-1L
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Assay:
    0.0998 - 0.1002 mol/l
  • Calibrated Against:
    sodium chloride

 

 

 :
 :169.87 g/mol
 :100 °C (1.013 hPa)
 :<0 °C
 :1.010 g/cm³ (20 °C)
 :Ambient
 :3264
 :8,III
 :GHS05
 :7761-88-8
 :AgNO₃
 :MFCD00003414