Order Entry
Czech Republic
ContactUsLinkComponent
Potassium hydroxide 1.0 M KOH (1.0 N) volumetric solution, Fluka™
Corrosive
Potassium hydroxide 1.0 M KOH (1.0 N) volumetric solution, Fluka™
Catalog # 319376-500ML
CAS Number:  
Corrosive
Potassium hydroxide 1.0 M KOH (1.0 N) volumetric solution, Fluka™
Catalog # 319376-500ML
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    complying
  • Coloration:
    complying
  • Clearness:
    complying
  • Assay titr.:
    0.950 - 1.050 mol/l
  • Traceable to NIST SRM:
    complying
  • BAM/EMPA reference material:
    complying
  • BAM/EMPA Certificate Number:
    complying
  • Carbonate (CO₃):
    Max. 0.01 mol/l

Specification Test Results

About this item

 :
 :56.11 g/mol
Density:1.05 g/cm³
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,II
 :GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553