

Nitrilotriacetic acid ≥99% for complexometry, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:139-13-9
:
Formula:C₆H₉NO₆
MDL Number:MFCD00004287
:191.14 g/mol
Merck Index:12,06675
Melting Pt:245 °C.242 °C
EINECS:205-355-7
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Assay:Min. 99 %
- Calcium (Ca):Max. 0.002 %
- Iron (Fe):Max. 0.0005 %
- Potassium (K):Max. 0.001 %
- Magnesium (Mg):Max. 0.001 %
- Sodium (Na):Max. 0.02 %
- Chloride (Cl):Max. 0.02 %
- Sulfate (SO₄):Max. 0.01 %
Specification Test Results
Frequently Bought Together