Order Entry
Czech Republic
ContactUsLinkComponent
Hydrochloric acid 6 M HCl (6N) volumetric solution, Fluka™
Hydrochloric acid 6 M HCl (6N) volumetric solution, Fluka™
Catalog # 31087-25L
CAS Number:  
Hydrochloric acid 6 M HCl (6N) volumetric solution, Fluka™
Catalog # 31087-25L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 L
  • Seq:
    5327

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

 :
Density:1.1 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1789
ADR:8,III
 :36.46 g/mol
 :GHS07
 :GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324