
Bromine (bromide-bromate) solution concentrate for dilution to 1L, 0.05 M Br2 volumetric solution, Fluka™
Catalog # 38040-1EA
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Ampoule
- Pk:1
- Seq:0001
Specifications
Specification Test Results
- Factor:0.998 - 1.002
Specification Test Results
About this item
:
:GHS08
Storage Temperature:Ambient
MDL Number:MFCD00010896