Order Entry
Czech Republic
ContactUsLinkComponent
Hydrochloric acid 6 M Reag. Ph. Eur.
Hydrochloric acid 6 M Reag. Ph. Eur.
Catalog # REGE3001500
Supplier:  REAGECON
CAS Number:  
Hydrochloric acid 6 M Reag. Ph. Eur.
Catalog # REGE3001500
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pack type:
    HDPE container
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :36.46 g/mol
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324