ContactUsLinkComponent
Amino acids analysis standards, AdvanceBio AAA
Amino acids analysis standards, AdvanceBio AAA
Catalog # HEWL5190-9426
Supplier:  Agilent
CAS Number:  
Amino acids analysis standards, AdvanceBio AAA
Catalog # HEWL5190-9426
Supplier:  Agilent
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    AdvanceBio amino acid reagents kit, 10 standards and reagents needed for amino acid analysis, consists of Borate buffer, FMOC, OPA, DTDPA, AA standards, 1-250 pmol/µL
Specifications
About this item
A kit with amino acid standards (17 components) at different concentrations together with reagents and buffers required for analysis.