ContactUsLinkComponent
Hydrophobic interaction chromatography columns, InfinityLab AdvanceBio
 :  Agilent
undefined
Hydrophobic interaction chromatography columns, InfinityLab AdvanceBio
 :  Agilent
  • Catalog No:
  • HEWL685975-908
  • HEWL681975-908
  • Max. pressure:
  • 400 bar
  • 400 bar
  • pH range:
  • 2,0 – 8,0
  • 2,0 – 8,0
  • Temperature limit:
  • 60 °C
  • 60 °C
Frequently Bought Together