ContactUsLinkComponent
Sulphate API agar medium
Sulphate API agar medium
Catalog # SIAL13005-500G-F
Supplier:  Merck
CAS Number:  
Sulphate API agar medium
Catalog # SIAL13005-500G-F
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    500 g
  • Description:
    Sulphate API agar
Specifications
About this item
Sulphate API medium is used for detection and estimation of sulphate reducing bacteria. These bacteria are able to grow and reduce sulphate to sulphide in this medium.
  • Supplement: 4 ml/l of sodium lactate
  • Final pH: 7,4 ±0,2 (25 °C)
Composition includes agar (14 g/l), ascorbic acid (0,1 g/l), dipotassium phosphate (0,01 g/l), ferrous ammonium sulphate (0,1 g/l), magnesium sulphate (0,2 g/l), sodium chloride (10 g/l) and yeast extract (1 g/l).