EDTA disodium salt solution with zinc complex added (solution B) for hardness of water determinations, for complexometry (1 ml = 1°dH/100 ml H₂O), Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
:
MDL Number:MFCD00147223
Boiling Pt:100 °C (1.013 hPa)
Density:1.000 g/cm³ (20 °C)
Storage Temperature:Ambient
Specification Test Results
- Factor:0.999 - 1.001
Specification Test Results
Frequently Bought Together