ContactUsLinkComponent
Desiccant pack, Whatman™
Desiccant pack, Whatman™
Catalog # WHAT10548239
undefined
Desiccant pack, Whatman™
Catalog # WHAT10548239
Specifications
  • Description:
    Desiccant pack
  • Size:
    1 g
  • Pk:
    100
Specifications
About this item
Desiccant pack ensure that DNA sampling cards remain dry during transport or storage.
  • High vapour transmission rate
  • Substrate is made with Tyvek® material
  • Non-dusting, strong and hard to tear