Orders LinkContactUsLinkComponent
AOCS-Tintometer colour reference standards, Supelco®
AOCS-Tintometer colour reference standards, Supelco®
Supplier:   Merck
AOCS-Tintometer colour reference standards, Supelco®
Supplier:  Merck
Certificates
AOCS-Tintometer colour reference standards, Supelco®
2 Options Available Below
Product Details & Documents
AOCS-Tintometer Colour Standard for routine calibration of colour measuring instruments and verification of test data. Process industries throughout the world use colorimeters for colour control of products such as edible and industrial oils, fuels, chemicals, pharmaceuticals, paints and coatings.
  • Certified under ISO 9001 Quality system.
  • Full Traceability to International Standards.
  • 500mL Pack Size supplied with MSDS
  • 12 months shelf life.
  • Tamper evident security packaging
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • SIAL134260-500ML
    AOCS-Tintometer Colour Reference Standard, 1.7R 12Y (5¼")
    AOCS-Tintometer Colour Reference Standard, 1.7R 12Y (5¼")
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 10 390,00 Kč
    10 390,00 Kč
    1 * 500 mL
       10 390,00 Kč
  • SIAL134270-500ML
    AOCS-Tintometer Colour Reference Standard, 3.0R 22Y (5¼")
    AOCS-Tintometer Colour Reference Standard, 3.0R 22Y (5¼")
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL