Immersion oil, Honeywell
Catalog # 10976-20ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle, Narrow Open
- Pk:20 mL
- Description:Immersion Oil
Specifications
About this item
UV-transparent, fluorescence-free immersion oil.
Specifications
Density (D 20/4): 1,075 to 1,104
Refractive index (n 20/D): 1,516 to 1,518
Fluorescent substances: complying