Order Entry
Czech Republic
ContactUsLinkComponent
5-(Ethylthio)-1H-tetrazole (ETT) 0.5M in acetonitrile, BioSyn™ activator reagent, for DNA/RNA synthesis, Burdick & Jackson™
Flammable
Health
Irritant
5-(Ethylthio)-1H-tetrazole (ETT) 0.5M in acetonitrile, BioSyn™ activator reagent, for DNA/RNA synthesis, Burdick & Jackson™
Catalog # HONCBR725-4X2.5GL
CAS Number:  
Flammable
Health
Irritant
5-(Ethylthio)-1H-tetrazole (ETT) 0.5M in acetonitrile, BioSyn™ activator reagent, for DNA/RNA synthesis, Burdick & Jackson™
Catalog # HONCBR725-4X2.5GL
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, GL45
  • Pk:
    10 L
  • Seq:
    5114
  • Pk Info:
    4x 2.5 L

Specifications

About this item

 :
Boiling Pt:82 °C (main component)
Melting Pt:-46 °C (main component)
Melting Pt:–46 °C (main component)
Density:0.809 g/cm³ (20 °C)
Flash Pt:13 °C
Storage Temperature:Ambient
UN:1648
ADR:3,II
 :130.17 g/mol
 :GHS02
 :GHS08
 :GHS07
CAS Number:89797-68-2
Formula:C₃H₆N₄S
MDL Number:MFCD00068733