Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Pepsin
Pepsin
Pepsin
176408.1214
:
PanReac AppliChem
:
Pepsin
176408.1214
:
PanReac AppliChem
:
Pk:
5 L
CAS No.:
9001-75-6
Storage conditions:
Ambient
Form:
Liquid
Enzyme name:
Pepsin