ContactUsLinkComponent
Flexible coupling for stirrers
Flexible coupling for stirrers
Catalog # WITG1.445.500
Supplier:  witeg Labortechnik
undefined
Flexible coupling for stirrers
Catalog # WITG1.445.500
Supplier:  witeg Labortechnik
Specifications
  • Description:
    Flexible coupling for stirrers
  • Pk:
    1
  • Shaft Ø:
    8 mm
Specifications
About this item
This flexible coupling fix between motor and stirrer shaft.
  • Made of PTFE/polyamid