Order Entry
Czech Republic
ContactUsLinkComponent
Ethidium Homodimer III (EthD-III) 1 mM in DMSO used in nuclear staining
Ethidium Homodimer III (EthD-III) 1 mM in DMSO used in nuclear staining
Catalog # 40051.
Supplier:  Biotium
CAS Number:  
undefined
Ethidium Homodimer III (EthD-III) 1 mM in DMSO used in nuclear staining
Catalog # 40051.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    200 µl
  • Seq:
    1358

Specifications

About this item

 :
 :~1000 g/mol
Storage Temperature:Refrigerator