Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethidium Homodimer III (EthD-III)
Ethidium Homodimer III (EthD-III) used in nuclear staining
Ethidium Homodimer III (EthD-III) used in nuclear staining
40050.
:
Biotium
:
Ethidium Homodimer III (EthD-III) used in nuclear staining
40050.
:
Biotium
:
Pk:
1 mg
Seq:
2070
:
:
~1000 g/mol
:
Refrigerator