Indo-1 pentasodium salt ≥95% (by HPLC) fluorescent indicator, for calcium determination
Catalog # 50042.
Supplier: Biotium
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:759.5 g/mol
:132319-56-3
Formula:C₃₂H₂₆N₃Na₅O₁₂