Order Entry
Czech Republic
ContactUsLinkComponent
FURA-2 pentapotassium salt ≥95% (by HPLC) fluorescent indicator, for calcium determination
FURA-2 pentapotassium salt ≥95% (by HPLC) fluorescent indicator, for calcium determination
Catalog # 50031.
Supplier:  Biotium
CAS Number:  
FURA-2 pentapotassium salt ≥95% (by HPLC) fluorescent indicator, for calcium determination
Catalog # 50031.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :832 g/mol
Storage Temperature:Refrigerator
CAS Number:113694-64-7
Formula:C₂₉H₂₂K₅N₃O₁₄