Order Entry
Czech Republic
ContactUsLinkComponent
FLUO 3/AM 1 mM in anhydrous DMSO fluorescent indicator, for calcium determination
FLUO 3/AM 1 mM in anhydrous DMSO fluorescent indicator, for calcium determination
Catalog # 50015.
Supplier:  Biotium
CAS Number:  
FLUO 3/AM 1 mM in anhydrous DMSO fluorescent indicator, for calcium determination
Catalog # 50015.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :1129.85 g/mol
Storage Temperature:Freezer
CAS Number:121714-22-5
Formula:C₅₁H₅₀Cl₂N₂O₂₃