FLUO-3 pentasodium salt ≥95% (by HPLC) fluorescent indicator, for calcium determination
Catalog # 50012.
Supplier: Biotium
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:879 g/mol
Storage Temperature:Refrigerator
CAS Number:134907-84-9
Formula:C₃₆H₂₅Cl₂Na₅N₂O₁₃