Order Entry
Czech Republic
ContactUsLinkComponent
Pluronic® F-127 10% in water
Pluronic® F-127 10% in water
Catalog # 59005.
Supplier:  Biotium
CAS Number:  
Pluronic® F-127 10% in water
Catalog # 59005.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    30 mL
  • Seq:
    2345

Specifications

About this item

 :
CAS Number:9003-11-6
Formula:HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)aH
MDL Number:MFCD00082049