Order Entry
Czech Republic
ContactUsLinkComponent
5,5'-difluoro BAPTA tetrapotassium salt ≥95% (by TLC) ion chelator, for calcium determination
5,5'-difluoro BAPTA tetrapotassium salt ≥95% (by TLC) ion chelator, for calcium determination
Catalog # 50006.
Supplier:  Biotium
CAS Number:  
5,5'-difluoro BAPTA tetrapotassium salt ≥95% (by TLC) ion chelator, for calcium determination
Catalog # 50006.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :665 g/mol
Storage Temperature:Refrigerator
CAS Number:152290-47-6
Formula:C₂₂H₁₈F₂K₄N₂O₁₀