Order Entry
Czech Republic
ContactUsLinkComponent
 
5,5'-difluoro BAPTA AM ester ≥95% (by TLC) ion chelator, for calcium determination
Catalog # 50005.
Supplier:  Biotium
CAS Number:  
5,5'-difluoro BAPTA AM ester ≥95% (by TLC) ion chelator, for calcium determination
Catalog # 50005.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :801 g/mol
Storage Temperature:Freezer
CAS Number:156027-00-8
Formula:C₃₄H₃₈F₂N₂O₁₈