Order Entry
Czech Republic
ContactUsLinkComponent
Trace Metals 1 in Seawater - Whole Volume, (CRM), Supelco®
Trace Metals 1 in Seawater - Whole Volume, (CRM), Supelco®
Catalog # QC3163-500ML
Supplier:  Merck
CAS Number:  
Trace Metals 1 in Seawater - Whole Volume, (CRM), Supelco®
Catalog # QC3163-500ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Description:
    Trace Metals 1 in Seawater - Whole Volume
  • Type:
    Certified reference material

Specifications

About this item