Order Entry
Czech Republic
ContactUsLinkComponent
Total Residual Chlorine-WP, (CRM), Supelco®
Total Residual Chlorine-WP, (CRM), Supelco®
Catalog # QC1065-2ML
Supplier:  Merck
CAS Number:  
Total Residual Chlorine-WP, (CRM), Supelco®
Catalog # QC1065-2ML
Supplier:  Merck
CAS Number:  

Specifications

  • Matrix:
    H₂O
  • Pk:
    2 mL
  • Description:
    Total Residual Chlorine-WP

Specifications

About this item

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.