Přeskočit na Záhlaví
Přeskočit na Hlavní obsah
Přeskočit na Patička
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Domů
Standards
Environmental Analysis Standards
Phenolics c/V sample 2, (CRM), Supelco®
Phenolics c/V sample 2, (CRM), Supelco®
Phenolics c/V sample 2, (CRM), Supelco®
Catalog # QC1167-2ML
Supplier:
Merck
CAS Number:
Expand image
Phenolics c/V sample 2, (CRM), Supelco®
Catalog # QC1167-2ML
Supplier:
Merck
CAS Number:
Specifications
Pk:
2 mL
Description:
Phenolics C/V Sample 2
Type:
Certified reference material
Specifications
About this item
Product Details
Product Details