0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Standards
Environmental Analysis Standards
Nonionic surfactants, (CRM), Supelco®
Nonionic surfactants, (CRM), Supelco®
Catalog # QC1197-20ML
Supplier:
Merck
CAS Number:
Expand image
Nonionic surfactants, (CRM), Supelco®
Catalog # QC1197-20ML
Supplier:
Merck
CAS Number:
Specifications
Pk:
20 mL
Description:
Nonionic Surfactants
Type:
Certified reference material
Specifications
About this item
Product Details
Product Details